2-Iodo-4-nitrophenol 98%

Supplier: Ambeed
Danger

A197821-100MG A197821-1G A197821-250MG A197821-5G
77097-304EA 13 USD
77097-304 77097-306 77097-308 77097-310
2-Iodo-4-nitrophenol 98%
2-Iodo-4-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄INO₃
MDL Number: MFCD01165664
CAS Number: 89487-91-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR