1-Acetyl-5-indolinesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 1-acetyl-2,3-dihydro-1H-indole-5-sulphonyl chloride, n-acetylindoline-5-sulphonyl chloride, 1-acetylindoline-5-sulphonyl chloride

A196720-1G A196720-250MG A196720-25G
77097-270EA 33 USD
77097-270 77097-272 77097-274
1-Acetyl-5-indolinesulfonyl chloride 98%
1-Acetyl-5-indolinesulfonyl chloride

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClNO₃S
MDL Number: MFCD07368558
CAS Number: 52206-05-0
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR