PF-06840003 98%

Supplier: Ambeed
Warning

Synonyms: 3‑(5-Fluoro‑1H‑indol-3-yl)pyrrolidine-2,5-dione, 3-(5-Fluoro-1H-indol-3-yl)pyrrolidine-2,5-dione, EOS 200271

A196131-10MG A196131-1MG A196131-50MG A196131-5MG
77097-252EA 92 USD
77097-252 77097-254 77097-256 77097-258
PF-06840003 98%
PF-06840003

Precaución: for Research Use Only

Formula: C₁₂H₉FN₂O₂
MW: 232.21 g/mol
MDL Number: MFCD25121820
CAS Number: 198474-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR