2-Methyl-2-(quinolin-6-yl)propanoic acid 95%

Supplier: Ambeed
Warning

A195883-100MG A195883-250MG
77097-248EA 94 USD
77097-248 77097-250
2-Methyl-2-(quinolin-6-yl)propanoic acid 95%
2-Methyl-2-(quinolin-6-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₂
MW: 215.25 g/mol
MDL Number: MFCD18260330
CAS Number: 1022283-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR