Quinoline-4-thiol 97%

Supplier: Ambeed
Warning

A195466-100MG A195466-250MG
77097-236EA 108.22 USD
77097-236 77097-238
Quinoline-4-thiol 97%
Quinoline-4-thiol

Precaución: for Research Use Only

Formula: C₉H₇NS
MDL Number: MFCD01861879
CAS Number: 51812-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR