N-Acetyl-L-tyrosine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Acetyl-L-Tyrosine, (S)-2-Acetamido-3-(4-hydroxyphenyl)propanoic acid, Ac-Tyr-OH
A195457-1000G
A195457-100G
A195457-10G
A195457-25G
A195457-500G
A195457-5G
77097-224EA
121
USD
77097-224
77097-226
77097-228
77097-230
77097-232
77097-234
N-Acetyl-L-tyrosine 98%
N-Acetyl-L-tyrosine
Precaución: for Research Use Only
Formula:
C₁₁H₁₃NO₄ MW: 223.23 g/mol |
MDL Number:
MFCD00037190 CAS Number: 537-55-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...