(S)-Benzyl 2-amino-4-methylpentanoate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-Leu-OBzl·HCl

A195042-10G A195042-1G A195042-250MG A195042-25G A195042-5G A195042-500G A195042-100G
77097-204EA 22.28 USD
77097-204 77097-206 77097-208 77097-210 77097-212 77685-482 77685-481
(S)-Benzyl 2-amino-4-methylpentanoate hydrochloride 97%
(S)-Benzyl 2-amino-4-methylpentanoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₂₀ClNO₂
MW: 257.76 g/mol
MDL Number: MFCD00034855
CAS Number: 2462-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR