(S)-1-(4-Bromophenyl)-2,2,2-trifluoroethanol 98%

Supplier: Ambeed
Warning

A194709-100MG A194709-1G A194709-250MG A194709-5G
77097-196EA 18.04 USD
77097-196 77097-198 77097-200 77097-202
(S)-1-(4-Bromophenyl)-2,2,2-trifluoroethanol 98%
(S)-1-(4-Bromophenyl)-2,2,2-trifluoroethanol

Precaución: for Research Use Only

Formula: C₈H₆BrF₃O
MW: 255.03 g/mol
MDL Number: MFCD14707649
CAS Number: 80418-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR