Methyl 4-((tert-butoxycarbonyl)amino)butanoate 95%

Supplier: Ambeed
Warning

A194669-1G A194669-250MG A194669-5G
77097-192EA 40.32 USD
77097-192 77097-194 77685-479
Methyl 4-((tert-butoxycarbonyl)amino)butanoate 95%
Methyl 4-((tert-butoxycarbonyl)amino)butanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
MW: 217.26 g/mol
MDL Number: MFCD18206247
CAS Number: 85909-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR