2-Fluorostyrene ≥95% stabilized

Supplier: Ambeed
Danger

Synonyms: 1-Fluoro-2-vinylbenzene

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A194083-5G A194083-1G A194083-100MG
77097-184EA 79.57 USD
77097-184 77567-810 77685-474
2-Fluorostyrene ≥95% stabilized
2-Fluorostyrene

Precaución: For research use only.

Formula: C₈H₇F
MW: 122.14 g/mol
MDL Number: MFCD00013550
CAS Number: 394-46-7
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR