tert-Butyl-6-formyl-1H-indole-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 6-formyl-1H-indole-1-carboxylate

A194040-100MG A194040-250MG A194040-5G A194040-1G
77097-178EA 32.89 USD
77097-178 77097-180 77097-182 77685-471
tert-Butyl-6-formyl-1H-indole-1-carboxylate 98%
tert-Butyl-6-formyl-1H-indole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO₃
MW: 245.28 g/mol
MDL Number: MFCD11877853
CAS Number: 127956-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR