Boc-L-Val-NH2 97%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl (1-amino-3-methyl-1-oxobutan-2-yl)carbamate, Boc-Val-NH2, Boc-Val-NH₂

A193605-1G A193605-250MG A193605-5G
77097-164EA 13 USD
77097-164 77097-166 77097-168
Boc-L-Val-NH2 97%
Boc-L-Val-NH2

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₃
MW: 216.28 g/mol
MDL Number: MFCD00190841
CAS Number: 35150-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR