4'-Methoxybiphenyl-4-ylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Methoxyphenyl)benzeneboronic acid, 4-(4-Methoxyphenyl)phenylboronic acid, (4'-Methoxy-[1,1'-biphenyl]-4-yl)boronic acid

A192286-1G A192286-250MG A192286-5G
77097-152EA 48.81 USD
77097-152 77097-154 77097-156
4'-Methoxybiphenyl-4-ylboronic acid 98%
4'-Methoxybiphenyl-4-ylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₃BO₃
MW: 228.0515 g/mol
MDL Number: MFCD04039030
CAS Number: 156642-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR