Ethyl 5-chloro-2-iodobenzoate 95%

Supplier: Ambeed
Warning

A191251-100MG A191251-10G A191251-1G A191251-250MG A191251-5G
77097-138EA 14.86 USD
77097-138 77097-140 77097-142 77097-144 77097-146
Ethyl 5-chloro-2-iodobenzoate 95%
Ethyl 5-chloro-2-iodobenzoate

Precaución: for Research Use Only

Formula: C₉H₈ClIO₂
MW: 310.51 g/mol
MDL Number: MFCD18396098
CAS Number: 1012882-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR