1-Bromo-4-(S-methylsulfonimidoyl)benzene 95%

Supplier: Ambeed
Warning

A190872-100MG A190872-250MG
77097-136EA 114 USD
77097-136 77685-455
1-Bromo-4-(S-methylsulfonimidoyl)benzene 95%
1-Bromo-4-(S-methylsulfonimidoyl)benzene

Precaución: for Research Use Only

Formula: C₇H₈BrNOS
MW: 234.11 g/mol
MDL Number: MFCD28991859
CAS Number: 145026-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR