N-Carbobenzoxyhydroxylamine 98%

Supplier: Ambeed
Warning

Synonyms: N-Hydroxycarbamic acid Benzyl Ester, N-Cbz-hydroxylamine, Benzyl N-Hydroxycarbamate, Benzylhydroxycarbamate

A190348-10G A190348-25G A190348-5G
77097-124EA 6 USD
77097-124 77097-126 77097-128
N-Carbobenzoxyhydroxylamine 98%
N-Carbobenzoxyhydroxylamine

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MDL Number: MFCD00010642
CAS Number: 3426-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR