Boc-2-fluoro-DL-phenylglycine 96%

Supplier: Ambeed
Warning

Synonyms: 2-((tert-Butoxycarbonyl)amino)-2-(2-fluorophenyl)acetic acid

A190107-100MG A190107-1G A190107-250MG
77097-116EA 37.14 USD
77097-116 77097-118 77097-120
Boc-2-fluoro-DL-phenylglycine 96%
Boc-2-fluoro-DL-phenylglycine

Precaución: for Research Use Only

Formula: C₁₃H₁₆FNO₄
MW: 269.28 g/mol
MDL Number: MFCD02682469
CAS Number: 161330-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR