(R)-1-Boc-2-(Hydroxymethyl)piperazine 97%
Supplier: Ambeed
|
Warning
|
A189815-100MG
A189815-1G
A189815-250MG
A189815-5G
A189815-10G
A189815-25G
77097-108EA
9
USD
77097-108
77097-110
77097-112
77097-114
77901-987
77901-045
(R)-1-Boc-2-(Hydroxymethyl)piperazine 97%
(R)-1-Boc-2-(Hydroxymethyl)piperazine
Precaución: for Research Use Only
Formula:
C₁₀H₂₀N₂O₃ |
MDL Number:
MFCD07772092 CAS Number: 169448-87-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...