2-Chloronicotinamide 98%

Supplier: Ambeed
Warning

A188695-100G A188695-10G
77097-076EA 76.39 USD
77097-076 77097-078
2-Chloronicotinamide 98%
2-Chloronicotinamide

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O
MW: 156.57 g/mol
MDL Number: MFCD00006237
CAS Number: 10366-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR