tert-Butyl (4-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate 98%

Supplier: Ambeed
Warning

Synonyms: 5-BOC-Amino-2-chlorophenylboronic acid pinacol ester

A188018-100MG A188018-1G A188018-250MG
77097-068EA 37.14 USD
77097-068 77097-070 77097-072
tert-Butyl (4-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate 98%
tert-Butyl (4-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₇H₂₅BClNO₄
MW: 353.65 g/mol
MDL Number: MFCD23379564
CAS Number: 1080573-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR