Bis[2-[(oxo)diphenylphosphino]phenyl] ether 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(Oxybis(2,1-phenylene))bis(diphenylphosphine oxide), DPEPO
A187345-10G
A187345-1G
A187345-250MG
A187345-5G
77097-050EA
5.31
USD
77097-050
77097-052
77097-054
77097-056
77097-058
Bis[2-[(oxo)diphenylphosphino]phenyl] ether 95%
Bis[2-[(oxo)diphenylphosphino]phenyl] ether
Precaución: for Research Use Only
Formula:
C₃₆H₂₈O₃P₂ |
MDL Number:
MFCD28968111 CAS Number: 808142-23-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...