2,4,5-Trichlorophenylboronic acid 97%

Supplier: Ambeed
Warning

A187086-100MG A187086-1G A187086-250MG A187086-5G
77097-042EA 32 USD
77097-042 77097-044 77097-046 77097-048
2,4,5-Trichlorophenylboronic acid 97%
2,4,5-Trichlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BCl₃O₂
MDL Number: MFCD09056706
CAS Number: 220210-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR