1-(3-Bromophenyl)pyrrolidine 98%

Supplier: Ambeed
Warning

A185871-1G A185871-250MG
77097-010EA 37.14 USD
77097-010 77097-012
1-(3-Bromophenyl)pyrrolidine 98%
1-(3-Bromophenyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrN
MW: 226.1130 g/mol
MDL Number: MFCD04112531
CAS Number: 219928-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR