2,6-Dimethoxynicotinic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dimethoxypyridine-3-carboxylic acid

A185410-100MG A185410-1G A185410-250MG A185410-5G
77096-996EA 16.98 USD
77096-996 77096-998 77097-000 77097-002
2,6-Dimethoxynicotinic acid 95%
2,6-Dimethoxynicotinic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₄
MW: 183.17 g/mol
MDL Number: MFCD00075582
CAS Number: 16727-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR