1-(3-Bromo-4-hydroxyphenyl)ethanone 97%

Supplier: Ambeed
Warning

Synonyms: 3'-Bromo-4'-hydroxyacetophenone, 3-Bromo-4-hydroxyacetophenone

A185181-100MG A185181-10G A185181-250MG A185181-5G
77096-988EA 26.53 USD
77096-988 77096-990 77096-992 77096-994
1-(3-Bromo-4-hydroxyphenyl)ethanone 97%
1-(3-Bromo-4-hydroxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
MDL Number: MFCD03208974
CAS Number: 1836-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR