N-Bocamino-cyano-acetic acid ethyl ester 95%

Supplier: Ambeed
Warning

A184766-100MG A184766-1G A184766-250MG A184766-5G
77096-978EA 44 USD
77096-978 77096-980 77096-982 77685-432
N-Bocamino-cyano-acetic acid ethyl ester 95%
N-Bocamino-cyano-acetic acid ethyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₄
MDL Number: MFCD11616708
CAS Number: 865370-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR