2-Nitro-5-(trifluoromethyl)benzonitrile 98%

Supplier: Ambeed
Warning

A184759-1G A184759-250MG
77096-974EA 17 USD
77096-974 77096-976
2-Nitro-5-(trifluoromethyl)benzonitrile 98%
2-Nitro-5-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₃F₃N₂O₂
MDL Number: MFCD09264505
CAS Number: 16499-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR