2-Bromo-5-nitro-3-picoline 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-3-methyl-5-nitropyridine

A184492-1G A184492-25G
77096-956EA 11 USD
77096-956 77096-958
2-Bromo-5-nitro-3-picoline 97%
2-Bromo-5-nitro-3-picoline

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₂
MW: 217.02 g/mol
MDL Number: MFCD03095065
CAS Number: 23132-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR