(5-Bromopyridin-3-yl)methanol hydrochloride 95+%

Supplier: Ambeed
Warning

A182614-10G A182614-250MG A182614-5G
77096-924EA 325.7 USD
77096-924 77096-926 77096-928
(5-Bromopyridin-3-yl)methanol hydrochloride 95+%
(5-Bromopyridin-3-yl)methanol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₇BrClNO
MW: 224.48 g/mol
MDL Number: MFCD11836367
CAS Number: 22620-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR