4-(Methylsulfonyl)benzonitrile 98%

Supplier: Ambeed
Warning

A182506-100G A182506-10G A182506-1G A182506-25G A182506-5G
77096-914EA 127 USD
77096-914 77096-916 77096-918 77096-920 77096-922
4-(Methylsulfonyl)benzonitrile 98%
4-(Methylsulphonyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₇NO₂S
MDL Number: MFCD00216489
CAS Number: 22821-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR