Diethyl 2,2'-dimethylbiphenyl-4,4'-dicarboxylate 95%

Supplier: Ambeed
Warning

A182443-100MG A182443-250MG
77096-910EA 36.08 USD
77096-910 77096-912
Diethyl 2,2'-dimethylbiphenyl-4,4'-dicarboxylate 95%
Diethyl 2,2'-dimethylbiphenyl-4,4'-dicarboxylate

Precaución: for Research Use Only

Formula: C₂₀H₂₂O₄
MW: 326.39 g/mol
MDL Number: MFCD21333033
CAS Number: 855254-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR