Fmoc-Dap(Fmoc)-OH 95%

Supplier: Ambeed
Warning

Synonyms: Nα-Fmoc-3-(Fmoc-amino)-L-alanine, (S)-2,3-Bis((((9H-fluoren-9-yl)methoxy)carbonyl)amino)propanoic acid

A182063-250MG A182063-25G A182063-5G A182063-1G
77096-904EA 7.43 USD
77096-904 77096-906 77096-908 77611-774
Fmoc-Dap(Fmoc)-OH 95%
Fmoc-Dap(Fmoc)-OH

Precaución: for Research Use Only

Formula: C₃₃H₂₈N₂O₆
MW: 548.595 g/mol
MDL Number: MFCD00237005
CAS Number: 201473-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR