2-Chloro-5,7-difluoroquinoline-3-carbaldehyde 97%

Supplier: Ambeed
Warning

A181938-1G A181938-250MG
77096-900EA 170.81 USD
77096-900 77096-902
2-Chloro-5,7-difluoroquinoline-3-carbaldehyde 97%
2-Chloro-5,7-difluoroquinoline-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₄ClF₂NO
MW: 227.60 g/mol
MDL Number: MFCD26399738
CAS Number: 956631-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR