tert-Butyl (trans-2-hydroxycyclopentyl)carbamate 95%

Supplier: Ambeed
Warning

A181902-1G A181902-250MG A181902-5G
77096-894EA 58 USD
77096-894 77096-896 77096-898
tert-Butyl (trans-2-hydroxycyclopentyl)carbamate 95%
tert-Butyl (trans-2-hydroxycyclopentyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MDL Number: MFCD09261430
CAS Number: 155837-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR