Ethyl 6-bromo-5-methylimidazo[1,2-a]pyridine-2-carboxylate 97%

Supplier: Ambeed
Warning

A181891-1G A181891-250MG A181891-5G
77096-888EA 51.99 USD
77096-888 77096-890 77096-892
Ethyl 6-bromo-5-methylimidazo[1,2-a]pyridine-2-carboxylate 97%
Ethyl 6-bromo-5-methylimidazo[1,2-a]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrN₂O₂
MW: 283.12 g/mol
MDL Number: MFCD19105298
CAS Number: 859787-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR