Methyl 2-amino-2-phenylpropanoate hydrochloride 97%

Supplier: Ambeed
Warning

A181817-100MG A181817-250MG
77096-884EA 47 USD
77096-884 77096-886
Methyl 2-amino-2-phenylpropanoate hydrochloride 97%
Methyl 2-amino-2-phenylpropanoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClNO₂
MDL Number: MFCD16817459
CAS Number: 191796-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR