Piperazine-2,6-dione 95%

Supplier: Ambeed
Warning

A181548-1G A181548-250MG A181548-25G
77096-872EA 50 USD
77096-872 77096-874 77096-876
Piperazine-2,6-dione 95%
Piperazine-2,6-dione

Precaución: for Research Use Only

Formula: C4H6N2O2
MDL Number: MFCD11109779
CAS Number: 4774-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR