Z-Lys(Z)-ONp 98%

Supplier: Ambeed
Warning

A181141-10G A181141-25G A181141-5G
77096-854EA 45 USD
77096-854 77096-856 77096-858
Z-Lys(Z)-ONp 98%
Z-Lys(Z)-ONp

Precaución: for Research Use Only

Formula: C₂₈H₂₉N₃O₈
MDL Number: MFCD00024657
CAS Number: 21160-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR