(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(((allyloxy)carbonyl)amino)pentanoic acid 97%

Supplier: Ambeed
Warning

A181125-1G A181125-250MG A181125-5G
77096-848EA 40 USD
77096-848 77096-850 77096-852
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(((allyloxy)carbonyl)amino)pentanoic acid 97%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(((allyloxy)carbonyl)amino)pentanoic acid

Precaución: for Research Use Only

Formula: C₂₄H₂₆N₂O₆
MDL Number: MFCD00798638
CAS Number: 214750-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR