6-Bromo-2,3-difluoronitrobenzene 95%

Supplier: Ambeed
Warning

A180860-100MG A180860-1G A180860-250MG A180860-5G
77096-828EA 37 USD
77096-828 77096-830 77096-832 77096-834
6-Bromo-2,3-difluoronitrobenzene 95%
6-Bromo-2,3-difluoronitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrF₂NO₂
MDL Number: MFCD03840797
CAS Number: 884495-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR