4-Bromo-4'-chloro-1,1'-biphenyl 99%

Supplier: Ambeed
Warning

A180515-100G A180515-10G A180515-1G A180515-250MG A180515-25G A180515-5G
77096-810EA 411 USD
77096-810 77096-812 77096-814 77096-816 77096-818 77096-820
4-Bromo-4'-chloro-1,1'-biphenyl 99%
4-Bromo-4'-chloro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈BrCl
MDL Number: MFCD06201384
CAS Number: 23055-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR