3-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-[3-(pyridin-3-yl)phenyl]-1,3,2-dioxaborolane, 3-(3-Pyridyl)phenylboronic acid Pinacol ester

A179924-1G A179924-5G
77096-794EA 24.41 USD
77096-794 77096-796
3-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine 97%
3-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine

Precaución: for Research Use Only

Formula: C₁₇H₂₀BNO₂
MW: 281.16 g/mol
MDL Number: MFCD20923943
CAS Number: 939430-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR