Fmoc-D-Lys(Mtt)-OH 97%

Supplier: Ambeed
Warning

A179858-10G A179858-250MG A179858-5G A179858-25G
77096-786EA 270 USD
77096-786 77096-788 77096-790 77901-613
Fmoc-D-Lys(Mtt)-OH 97%
Fmoc-D-Lys(Mtt)-OH

Precaución: for Research Use Only

Formula: C₄₁H₄₀N₂O₄
MDL Number: MFCD26793593
CAS Number: 198544-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR