2-Amino-5-chloro-3-fluoropyridine 97%

Supplier: Ambeed
Warning

A178937-100G A178937-10G A178937-25G A178937-5G
77096-766EA 95 USD
77096-766 77096-768 77096-770 77096-772 77096-774
2-Amino-5-chloro-3-fluoropyridine 97%
2-Amino-5-chloro-3-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₄ClFN₂
MDL Number: MFCD09878434
CAS Number: 246847-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR