4-Fluoro-3-methylbenzene-1,2-diamine 97%

Supplier: Ambeed
Warning

A178781-100MG A178781-1G A178781-250MG
77096-760EA 77 USD
77096-760 77096-762 77096-764
4-Fluoro-3-methylbenzene-1,2-diamine 97%
4-Fluoro-3-methylbenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₇H₉FN₂
MDL Number: MFCD09026959
CAS Number: 485832-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR