2,4-Dihydroxy-3-nitroquinoline 98%

Supplier: Ambeed
Warning

A178153-1G A178153-25G A178153-5G
77096-748EA 13 USD
77096-748 77096-750 77096-752
2,4-Dihydroxy-3-nitroquinoline 98%
2,4-Dihydroxy-3-nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₆N₂O₄
MDL Number: MFCD01100327
CAS Number: 15151-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR