(Chloromethyl)triphenylphosphonium chloride 98%

Supplier: Ambeed
Warning

A178128-100G A178128-1G A178128-25G
77096-736EA 289 USD
77096-736 77096-738 77096-740
(Chloromethyl)triphenylphosphonium chloride 98%
(Chloromethyl)triphenylphosphonium chloride

Precaución: for Research Use Only

Formula: C₁₉H₁₇Cl₂P
MDL Number: MFCD00011867
CAS Number: 5293-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR