4-Bromo-8-trifluoromethylquinoline 95%

Supplier: Ambeed
Warning

A177974-100MG A177974-1G A177974-250MG A177974-5G
77096-716EA 14 USD
77096-716 77096-718 77096-720 77096-722
4-Bromo-8-trifluoromethylquinoline 95%
4-Bromo-8-trifluoromethylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₅BrF₃N
MDL Number: MFCD08063211
CAS Number: 260973-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR