4-Chloro-N-methoxy-N-methylpyridine-2-carboxamide 97%

Supplier: Ambeed
Warning

A177583-100G A177583-10G A177583-1G A177583-25G A177583-5G
77096-686EA 335.25 USD
77096-686 77096-688 77096-690 77096-692 77096-694
4-Chloro-N-methoxy-N-methylpyridine-2-carboxamide 97%
4-Chloro-N-methoxy-N-methylpyridine-2-carboxamide

Precaución: for Research Use Only

Formula: C₈H₉ClN₂O₂
MW: 200.62 g/mol
MDL Number: MFCD11975556
CAS Number: 757251-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR