Silibinin 98%

Supplier: Ambeed
Warning

Synonyms: Silybin A, (2R,3R)-3,5,7-Trihydroxy-2-((2R,3R)-3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)chroman-4-one

A177518-100MG A177518-10G A177518-1G A177518-25MG A177518-25G A177518-5G A177518-1MG
77096-678EA 9 USD
77096-678 77096-680 77096-682 77096-684 77611-724 77611-726 77685-399
Silibinin 98%
Silibinin

Precaución: for Research Use Only

Formula: C₂₅H₂₂O₁₀
MDL Number: MFCD03225424
CAS Number: 22888-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR